Anti-p-CaMKIV Rabbit Polyclonal Antibody [clone: T196/200]
Catalog # BWRLBS4300
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-CaMKIV
- Clonality:Polyclonal
- Clone:T196/200
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human CaMKIV around the phosphorylation site of Threonine 196/200.
p-CaMKIV (T196/200) pAb detects endogenous levels of human CaMKIV protein only when phosphorylated at Thr200. This antibody also detects mouse and rat CaMKIV protein when phosphorylated at the corresponding sites (Thr196).
Type: Primary
Antigen: p-CaMKIV
Clonality: Polyclonal
Clone: T196/200
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat