Anti-p-Na+/K+-ATPase alpha1 Rabbit Polyclonal Antibody [clone: S16]
Catalog # BWRLBS4259
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Na+/K+-ATPase alpha1
- Clonality:Polyclonal
- Clone:S16
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Na+/K+-ATPase alpha1 around the phosphorylation site of Serine 16.
p-Na+/K+-ATPase ?1 (S16) pAb detects endogenous levels of Na+/K+-ATPase ?1 protein only when phosphorylated at Ser16.
Type: Primary
Antigen: p-Na+/K+-ATPase alpha1
Clonality: Polyclonal
Clone: S16
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat