Anti-TWIK-1 Rabbit Polyclonal Antibody [clone: V322]
BWRLBS3370
- Antibody type:Primary
- Antigen symbol:TWIK-1
- Clonality:Polyclonal
- Clone:V322
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 301-350 of Human TWIK-1.
TWIK-1 (V322) pAb detects endogenous levels of TWIK-1 protein.
Type: Primary
Antigen: TWIK-1
Clonality: Polyclonal
Clone: V322
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat