Anti-Synuclein-alpha Rabbit Polyclonal Antibody [clone: E123]
Catalog # BWRLBS1596
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Synuclein-alpha
- Clonality:Polyclonal
- Clone:E123
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 100-150 of Human Synuclein-alpha.
SNCA (E123) pAb detects endogenous levels of SNCA protein.
Type: Primary
Antigen: Synuclein-alpha
Clonality: Polyclonal
Clone: E123
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat