Order Entry
Belgium
ContactUsLinkComponent
Anti-Na+ CP type alpha-pan Rabbit Polyclonal Antibody [clone: E1476]
Anti-Na+ CP type alpha-pan Rabbit Polyclonal Antibody [clone: E1476]
Catalog # BWRLBS3558
Anti-Na+ CP type alpha-pan Rabbit Polyclonal Antibody [clone: E1476]
Catalog # BWRLBS3558

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Na+ CP type alpha-pan
  • Clonality:
    Polyclonal
  • Clone:
    E1476
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 1442-1496 of Human Na+ CP type alpha-pan.

Na+ CP type ?-pan (E1476) pAb detects endogenous levels of Na+ CP type ?-pan protein.

Type: Primary
Antigen: Na+ CP type alpha-pan
Clonality: Polyclonal
Clone: E1476
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat