Anti-Neurexophilin-3 Rabbit Polyclonal Antibody [clone: L190]
Catalog # BWRLBS3891
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Neurexophilin-3
- Clonality:Polyclonal
- Clone:L190
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 161-210 of Human Neurexophilin-3.
Neurexophilin-3 (L190) pAb detects endogenous levels of Neurexophilin-3 protein.
Type: Primary
Antigen: Neurexophilin-3
Clonality: Polyclonal
Clone: L190
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat