ContactUsLinkComponent
Dissolved oxygen, reagent set
Catalog # HACH2351400
Supplier:  Hach
CAS Number:  
undefined
Dissolved oxygen, reagent set
Catalog # HACH2351400
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    Winkler
  • Pk:
    1 SET
  • Description:
    Dissolved oxygen reagent set, including burette
  • Environmentally Preferable:
  • Sample size:
    300 ml
  • Tests per kit:
    100
  • Range:
    1 - 10 mg/l O2
Specifications
About this item
For determination of dissolved oxygen by the Winkler method (azide modification) and buret titration.