Order Entry
Belgium
ContactUsLinkComponent
Orion Star™ A326 Multi-Parameter Meter (pH/RDO/DO), Handheld
Orion Star™ A326 Multi-Parameter Meter (pH/RDO/DO), Handheld
Supplier:  Thermo Orion
Certificates
undefined
Orion Star™ A326 Multi-Parameter Meter (pH/RDO/DO), Handheld
Supplier:  Thermo Orion

Specifications

  • Catalog No:
  • 665-0279
  • 665-0280
  • Model:
  • A326
  • A326
  • pH range:
  • –2,000...+20,000
  • –2,000...+20,000
  • pH resolution:
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • pH calibration:
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • mV range:
  • ±2000,0
  • ±2000,0
  • mV resolution:
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,2 mV or 0,05% of reading whichever is greater
  • ±0,2 mV or 0,05% of reading whichever is greater
  • DO concentration:
  • 0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
  • 0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
  • DO concentration resolution:
  • 0,1 / 0,01 mg/L
  • 0,1 / 0,01 mg/L
  • DO saturation:
  • 0,0 - 600,0 % (polarographic); 0,0 - 500,0 % (RDO)
  • 0,0 - 600,0 % (polarographic); 0,0 - 500,0 % (RDO)
  • DO accuracy:
  • Polarographic: ±0,2 mg/L or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/L (up to 8 mg/L), ±0,2 mg/L (8 - 20 mg/L), ±10% of reading (up to 50 mg/L)
  • Polarographic: ±0,2 mg/l or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/l (up to 8 mg/l), ±0,2 mg/l (8 - 20 mg/l), ±10% of reading (up to 50 mg/l)
  • DO calibration:
  • Water-saturated air, air saturated water, manual (Winkler) and zero point
  • Water-saturated air, air saturated water, manual (Winkler) and zero point
  • DO barometric pressure range:
  • Automatic or manual, 450 - 850 mmHg
  • Automatic or manual, 450 - 850 mmHg
  • DO resolution:
  • 0.1, 1 % saturation
  • 0.1, 1 % saturation
  • Temperature range:
  • –5,0...+105,0 °C
  • –5,0...+105,0 °C
  • Temperature resolution:
  • 0,1 °C, 0,1 °F
  • 0,1 °C, 0,1 °F
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • Temperature calibration:
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • Temperature compensation:
  • Automatic or manual
  • Automatic or manual
  • Pressure resolution:
  • 1 mm Hg
  • 1 mm Hg
  • Pressure accuracy:
  • 6 mm Hg
  • 6 mm Hg
  • Display:
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Battery type:
  • 4 AA (included)
  • 4 AA (included)
  • Battery life:
  • 800 hours
  • 800 hours
  • Power supply:
  • AC adapter or 4 AA batteries
  • AC adapter or 4 AA batteries
  • Compliance:
  • IP 67
  • IP 67
  • Data logging - log function:
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Data logging capacity:
  • 5000 with date and time stamp
  • 5000 with date and time stamp
  • Memory:
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Languages:
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • IP rating:
  • IP 67
  • IP 67
  • Connections:
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature)
  • Outputs:
  • RS232, USB
  • RS232, USB
  • Alarm:
  • High/low limit alarm, calibration due alarm
  • High/low limit alarm, calibration due alarm
  • No. of channels:
  • 2
  • 2
  • Calibration:
  • pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
  • pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
  • Calibration points:
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • W×D×H:
  • 240×100×65 mm
  • 240×100×65 mm
  • Weight:
  • 0,5 kg
  • 0,5 kg

Specifications

Frequently Bought Together