Order Entry
Belgium
ContactUsLinkComponent
Potassium carbonate 5 M in aqueous solution for analysis
Potassium carbonate 5 M in aqueous solution for analysis
Catalog # CAYM700517-5
Supplier:  Cayman Chemical
CAS Number:  
undefined
Potassium carbonate 5 M in aqueous solution for analysis
Catalog # CAYM700517-5
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    5 ml
  • Seq:
    2210

Specifications

About this item

 :
 :138.21 g/mol
CAS Number:584-08-7
Formula:K₂CO₃
MDL Number:MFCD00011382