Protein carbonyl colorimetric assay kit
CAYM10005020-96
:
- Pk:96 Tests
- Description:Protein Carbonyl Colorimetric Assay Kit
- Storage temperature:-20°C
- Tests per kit:96
The Protein Carbonyl Colorimetric Assay Kit is for the measurement of oxidised proteins. Protein samples are derivatised by making use of the reaction between 2,4-dinitrophenylhydrazine and protein carbonyls.
- Measure protein carbonyl content in plasma, serum, urine, tissue homogenates and cell lysates
- Assay 24 samples in duplicate
- Plate-based colorimetric measurement (360 to 385 nm)