About this item
Supracap™ 100 depth filter capsules with PP housing are scalable capsules with Seitz® high performance depth filter media. They are suitable for process development and pilot scale applications.
- Seamless linear scalability
- Completely disposable
- Encapsulated design reduces operator exposure to potential biohazards
Low hold-up volume allows for increased product recovery and requires low post-use rinse volumes.
Applications include biopharmaceuticals, mammalian cell cultures, therapeutic protein, vaccines, blood plasma proteins and serum, yeast, E.coli lysates and refolds.
The Seitz® Bio 20 media grade is made of highly purified natural and modified celluloses and is free from inorganic materials. Seitz® Bio 20 media is ideal for ion sensitive products such as therapeutic proteins, dialysis solutions, and protein-based diagnostics.
Seitz® P-series media are comprised of cellulose fibers, filter aids (diatomaceous earth and perlite), and resins. This grade is recommended for coarse filtration in the biotechnological and pharmaceutical industries.
Seitz® HP-series media are feature two distinct layers of Seitz® P-series depth filter sheets that feature a more permeable layer followed by a less permeable layer (PDD1, PDE2, PDH4 and PDK5). Seitz® HP-series media performs exceptionally well in low viability, high solids-containing applications, and with wider distribution of particle sizes in biotech applications.
Specifications
- Catalog No:
- PALLNP6P7001
- PALLNP6PDK56
- PALLNP5LPDD16
- PALLNP5LPDE26
- PALLNP5LPDH41
- PALLNP6P2501
- PALLNP6P2506
- PALLNP5LB0201
- PALLNP6PDK51
- PALLNP6P2001
- PALLNP6P2006
- PALLNP5LPDD11
- PALLNP5LP1006
- PALLNP5LP2506
- PALLNP6PDP86
- PALLNP6PDP81
- PALLNP5LP7001
- PALLNP5LP2006
- PALLNP5LP2501
- 514-1411
- PALLNP5LPDE21
- PALLNP5LP7006
- PALLNP5LP9001
- PALLNP5LP9006
- PALLNP6P7006
- PALLNP6P1006
- PALLNP5LPDK51
- PALLNP5LPDK56
- PALLNP5LP1001
- PALLNP6B0201
- PALLNP6PDD11
- PALLNP5LB0206
- PALLNP6PDD16
- PALLNP5LPDH46
- PALLNP6PDH46
- PALLNP5LP2001
- PALLNP6PDE21
- Ø×L:
- 154×397 mm
- 154×263 mm
- 154×263 mm
- 154×213 mm
- 154×335 mm
- 154×397 mm
- 154×213 mm
- 154×335 mm
- 154×335 mm
- 154×397 mm
- 154×213 mm
- 154×263 mm
- 154×263 mm
- 154×213 mm
- 154×263 mm
- 154×213 mm
- 154×263 mm
- 154×213 mm
- 154×263 mm
- 154×397 mm
- 154×213 mm
- 154×263 mm
- 154×213 mm
- 154×335 mm
- 154×335 mm
- 154×263 mm
- 154×397 mm
- 154×263 mm
- 154×397 mm
- 154×213 mm
- 154×335 mm
- Max. temperature:
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- 40 °C
- Max. pressure:
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
- 3 bar
Specifications
Related Information
Pall Laboratory Supracap™ Depth Filter Capsules Application Note.pdf