Orders LinkContactUsLinkComponent
Pads, D-Formalizer®
Pads, D-Formalizer®
Supplier:   LEICA HISTOLOGY
Pads, D-Formalizer®
Supplier:  LEICA HISTOLOGY
Certificates
Pads, D-Formalizer®
Pads, D-Formalizer®
2 Options Available Below
Product Details & Documents
These D-Formalizer® pads reduces formaldehyde exposure by absorbing and neutralising any excess formalin.
  • Highly durable
  • Can be used to line shelves
  • Highly contrasting blue surface
The durable blue surface of the pads provides contrast to even small pieces of tissue. The Pads are also useful for soaking up leakage and de-odorising vapors in other applications. They can be used to line shelves which hold specimen containers that may seep formaldehyde around the lid, or to line transport boxes where leaks may occur.
Product Family Options
Product Information
  • L×W
    Availability
    Price
  • LEIH38007530810
    D-Formalizer® Pads
    D-Formalizer® Pads
    203×254 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 168,00 €
    168,00 €
    1 * 25 ST
       168,00 €
  • LEIH38007531114
    D-Formalizer® Pads
    D-Formalizer® Pads
    280×356 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST