Orders LinkContactUsLinkComponent
StarFish multiple experiment workstation
StarFish multiple experiment workstation
StarFish multiple experiment workstation
Certificates
StarFish multiple experiment workstation
StarFish multiple experiment workstation
8 Options Available Below
Product Details & Documents
Converts a magnetic stirrer into a multiple experiment workstation, improve the productivity of your laboratory; use a single stirring hotplate to do multiple experiments. Perfect for simple heating/stirring experiments or more complex applications such as synthesis, concentrations, extractions and distillations. Accepts a wide range of standard laboratory glassware including vials, test tubes, beakers and round bottom flasks, a range of inserts are available for small vials up to 250 ml round bottom flasks.
  • Powerful stirring and heating of all positions simultaneously
  • Truly versatile, mix and match up to five different PolyBlocks of the same or different size, or use a single MonoBlock with multiple wells all the same size
  • Connectors feature leak-proof shut-off valves
Universal five-way telescopic clamps, the StarFish clamp allows glassware of virtually any size to be used and comes with a choice of silicone rubber or Velcro support straps. Each clamp features 5 telescopic arms which can be extended and locked in place to suit your needs. Using different straps on separate clamps means the flask neck can be gripped with the silicone rubber strap for lifting, whilst the condenser above slides through the Velcro strap.
Water Distribution Manifold allows coolant from a single source to be evenly distributed to up to five condensers and then the flow re-combined to one outlet pipe. Two manifolds are used in each set-up, one to distribute water to the condensers and one to collect coolant for recirculation or to drain.
Gas/Vacuum Distribution Manifold allows gas or a vacuum from a single source to be evenly distributed to up to five positions or vessels. (Does not control or regulate gas/vacuum flow).
Product Family Options
MonoBlocks
Product Information
  • Availability
    Price
  • 442-0604
    MonoBlock for 5×250 ml flasks
    MonoBlock for 5×250 ml flasks
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 674,00 €
    674,00 €
    1 * 1 ST
       674,00 €
Base plates
Product Information
  • Availability
    Price
  • 442-0644
    Starfish base-plate handles (pair)
    Starfish base-plate handles (pair)
    Log in for availability
    Log in for availability
    1 * 1 PAIRE
    1 * 1 PAIRE - 601,00 €
    601,00 €
    1 * 1 PAIRE
       601,00 €
PolyBlocks
Product Information
  • Availability
    Price
  • 442-0619
    PolyBlock for 3×28 mm vials
    PolyBlock for 3×28 mm vials
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 245,00 €
    245,00 €
    1 * 1 ST
       245,00 €
Inserts
Product Information
  • Availability
    Price
  • 442-0626
    Flask insert 50 ml
    Flask insert 50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 111,00 €
    111,00 €
    1 * 1 ST
       111,00 €
  • 442-0624
    Flask insert 150 ml
    Flask insert 150 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 111,00 €
    111,00 €
    1 * 1 ST
       111,00 €
StarFish accessories
Product Information
  • Availability
    Price
  • 442-0649
    Support rod 650 mm
    Support rod 650 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 106,00 €
    106,00 €
    1 * 1 ST
       106,00 €
  • 442-0650
    Split support rod 650 mm
    Split support rod 650 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €
  • 442-0645
    Five way clamp (Velcro)
    Five way clamp (Velcro)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 548,00 €
    548,00 €
    1 * 1 ST
       548,00 €