Order Entry
Belgium
ContactUsLinkComponent
Genomic DNA extraction from dried blood spots, Extracta™ DBS
Genomic DNA extraction from dried blood spots, Extracta™ DBS
Supplier:  Quantabio
Certificates
undefined
Genomic DNA extraction from dried blood spots, Extracta™ DBS
Supplier:  Quantabio

Specifications

  • Method / Format:
    Extraction for direct PCR
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood (dried) on filter cards
  • Target molecule:
    DNA
  • Description:
    Extracta™ DBS

Specifications

Frequently Bought Together