Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Glucose hemolysing solution

Glucose hemolysing solution, 500 mL
Catalog # DIAS125809990338
Glucose hemolysing solution
Catalog # DIAS125809990338
CAS Number:  
Glucose hemolysing solution
Glucose hemolysing solution

Glucose hemolysing solution, 500 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents

For use with glucose Gluc-DH FS and glucose Gluc-DH FSC.

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Glucose hemolysing solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.