Orders LinkContactUsLinkComponent
Embedding system, Technovit® 7100
Embedding system, Technovit® 7100
Supplier:   HERAEUS KULZER
Embedding system, Technovit® 7100
Supplier:  HERAEUS KULZER
Certificates
Embedding system, Technovit® 7100
Embedding system, Technovit® 7100
5 Options Available Below
Product Details & Documents
Technovit 7100 is a plastic embedding system based on HEMA (2-hydroxyethyl methacrylate). The hydrophillic resin is used in medicine, botany, zoology and in the industry for embedding tissues for light microscope studies. The sections can be used for histological staining and enzyme detection.
  • Reproducibility and reliability of the embedding due to the constant, documented quality controls of the individual components
  • Low polymerisation temperature due to Teflon embedding forms
  • Equal hardening of the block, thus uniform and thinnest possible sections
  • In addition to routine staining, enzyme detection is also possible
  • Less toxic due to catalyst made of barbituric acid
  • Decalcification not necessary for haematological illiac crest biopsies
Technovit 7100 transparently polymerises. Uniform thin sections can be made out of the blocks with the rotation microtome. It is not necessary and also not possible to elute the plastic out of the block and the section.
Areas of applications for embeddings and sections include plastics, foils, paper, textiles, biological products and mounts and wires.
Delivery information: Contains a basic solution (500 ml), hardner 1 (5×1 g) and hardner 2 (1×40 ml).
Product Family Options
Product Information
  • Availability
    Price
  • HKUL64708955
    PE embedding mould, Ø 25 mm
    PE embedding mould, Ø 25 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 56,40 €
    56,40 €
    1 * 3 ST
       56,40 €
  • HKUL66009903
    Insert, for Ø 25 mm embedding mould
    Insert, for Ø 25 mm embedding mould
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 55,00 €
    55,00 €
    1 * 3 ST
       55,00 €
  • HKUL64708991
    Histoform S
    Histoform S
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 230,00 €
    1 230,00 €
    1 * 1 ST
       1 230,00 €
  • HKUL64708992
    Histoform Q
    Histoform Q
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HKUL64708996
    Blade holder, 17 cm
    Blade holder, 17 cm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST