Order Entry
Belgium
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Supplier:  MP Biomedicals
Certificates
undefined
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99.95%
  • FTIR:
    Conforms to Standard
  • pH (5% aq soln):
    10-11.5
  • OD 260 nm (5% aq soln):
    ≤0.1
  • Loss on Drying:
    ≤1%
  • Solubility (40% aq soln):
    Clear, Colorless Solution

Specification Test Results

Frequently Bought Together