Order Entry
Belgium
ContactUsLinkComponent
Neuraminidase, MP Biomedicals
Neuraminidase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Neuraminidase, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9001-67-6
  • Storage conditions:
    +4 °C
  • Enzyme name:
    Neuraminidase

Specifications

Frequently Bought Together