Pig Trypsin (from Pancreas), MP Biomedicals
- Dry powder: stable
- Protease act: min 250,000 USP units/gm.
pH: ~ 8,0 (Optimum) (Lit.)
- CAS No.:9002-07-7
- Source:Pancreas
- Species:Pig
- Storage conditions:+4 °C
- Form:Off-white powder
- Enzyme name:Trypsin
- Enzyme activity:>250 USP u/mg
- Enzyme synonyms:Parenzymol, Parenzyme, Tryptar, Trypure, U-4858
- Molecular weight:24000
- pH:~8,0 (Optimum) (Lit.)
Frequently Bought Together