ContactUsLinkComponent
Lecithin (from soy bean), dark yellow powder
Lecithin (from soy bean), dark yellow powder
  • Identity Test:
    Passes
  • Source:
    Soybean (refined oil removed)
  • Moisture:
    ≤2%
  • Acetone Insolubles:
    ≥95%
Frequently Bought Together