
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥99% (by titrimetric analysis), white powder
:6976-37-0
:
:C₈H₁₉NO₅
:MFCD00002853
:209.24 g/mol
:101…106 °C
:GHS07
:Ambient
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- pH (1% aq soln):9-10
- Moisture:≤1%
Frequently Bought Together