Bovine alpha-Chymotripsin (from Pancreas), MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Preparation Method
Produced from 3× crystallised chymotrypsinogen
α-Chymotrypsin is used for treating pancreatic insufficiency and in traumatology.
Chymotrypsin preferentially catalyses the hydrolysis of peptide bonds involving L-isomers of tyrosine, phenylalanine, and tryptophan. It also readily acts upon amides and esters of susceptible amino acids. In addition to bonds involving aromatic amino acids, chymotrypsin catalyses at a high rate the hydrolysis of bonds of leucyl, methionyl, asparaginyl, and glutamyl residues. a-Chymotrypsin is a protein consisting of 241 amino acid residues. The molecule has three peptide chains: an A chain of 13 residues, a B chain of 131 residues, and a C chain of 97 residues.
Storage temperature: –20 °C
Specifications
- Source:Pancreas
- Species:Bovine
- Storage conditions:Store at +4 °C
- Form:White powder
- Enzyme name:alpha-Chymotripsin
- Enzyme synonyms:E.C. 3.4.21.1, α-Chymotripsin
- Concentration:Protein: ≥80%
Specifications
Frequently Bought Together