Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyamide
Polyamide
Polyamide
ICNA0209602
:
MP Biomedicals
:
Polyamide
ICNA0209602
:
MP Biomedicals
:
Pk:
250 g
Seq:
3545
:
:
0.2 g/mL (bulk density)
:
Ambient
:
MFCD00217747