Alumina B, activity super I, white to light pink free flowing powder, MP EcoChrom™ for dioxin analysis
Catalog # ICNA0204569
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Identity Test:Passes
- pH:9.3-10.1
- Activity acc to Brockmann:14 - 16 l/mm
- Loss of Ignition:0.6-0.8%
- Particle Size Distribution Fine Parts <63 µm:≤10%
- Particle Size Distribution Large Parts >200 µm:≤5%
Specification Test Results
About this item
:
:101.96 g/mol
Boiling Pt:∼2980 °C
Melting Pt:∼. 2072 °C
Density:3.94 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:1344-28-1
Formula:Al₂O₃
MDL Number:MFCD00003424