Silicon dioxide, Ecochrom™ 32-63 µm
2,5 kg
Catalog # ICNA0202827
Supplier: MP Biomedicals
Silicon dioxide, Ecochrom™ 32-63 µm
Catalog # ICNA0202827
Supplier: MP Biomedicals
CAS Number: 7631-86-9
Jump to:
Product Details & Documents
- Specific Surface Area: 500-600 m2/g
- Particle Size: 32-63 µm
- Weight: 0.4-0.5 g/ml
- pH: 7.0
- Mean Pore Size: 60Å
pH: 7
Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
- CAS Number:7631-86-9
- Formula:SiO₂
- MDL Number:MFCD00011232
- Molecular Weight:60.08 g/mol
- Density:∼2 g/cm³ (20 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Pk:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



