Orders LinkContactUsLinkComponent
Abbe refractometers, NAR
Supplier:   Atago
Abbe refractometers, NAR
Supplier:  Atago
Certificates
Abbe refractometers, NAR
5 Options Available Below
Product Details & Documents
The NAR Abbe refractometers can meet many of the requirements for routine measurements.
Single wavelengths and standard temperatures can be measured at extended ranges, as well as elevated temperatures, measurements of solid samples (films, plastic sheets).
Specifications for Product Family
Specifications
  • Catalog No:
  • ATAG1220
  • ATAG1230
  • ATAG1240
  • ATAG1211
  • ATAG1212
  • Model:
  • NAR-2T
  • NAR-3T
  • NAR-4T
  • NAR-1T (liquids)
  • NAR-1T (solids)
  • Light source:
  • LED
  • LED
  • LED
  • LED
  • LED
  • Temperature measurement:
  • 5...120 °C
  • 5...50 °C
  • 5...50 °C
  • 5...50 °C
  • 5...50 °C
  • Temperature sensor:
  • ±0,2 °C
  • ±0,2 °C
  • ±0,2 °C
  • ±0,2 °C
  • ±0,2 °C
  • Temperature sensor accuracy:
  • ±0,2/0,5 °C
  • ±0,2 °C
  • ±0,2 °C
  • ±0,2 °C
  • ±0,2 °C
Specifications
Product Family Options
Product Information
  • Model
    Range
    Refractive index
    Availability
    Price
  • NAR-2T
    0 − 95 °Bx
    1,30 − 1,70 nD
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 030,00 €
    5 030,00 €
    1 * 1 ST
       5 030,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ATAG1220
    Model
    NAR-2T
    Light source
    LED
    Temperature measurement
    5...120 °C
    Temperature sensor
    ±0,2 °C
    Temperature sensor accuracy
    ±0,2/0,5 °C
  • NAR-3T
    0 − 95 °Bx
    1,30 − 1,71 nD
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 450,00 €
    5 450,00 €
    1 * 1 ST
       5 450,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ATAG1230
    Model
    NAR-3T
    Light source
    LED
    Temperature measurement
    5...50 °C
    Temperature sensor
    ±0,2 °C
    Temperature sensor accuracy
    ±0,2 °C
  • NAR-4T
    0 − 95 °Bx
    1,47 − 1,87 nD
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 450,00 €
    4 450,00 €
    1 * 1 ST
       4 450,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ATAG1240
    Model
    NAR-4T
    Light source
    LED
    Temperature measurement
    5...50 °C
    Temperature sensor
    ±0,2 °C
    Temperature sensor accuracy
    ±0,2 °C
  • NAR-1T (liquids)
    0 − 95 °Bx
    1,30 − 1,70 nD
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 920,00 €
    3 920,00 €
    1 * 1 ST
       3 920,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ATAG1211
    Model
    NAR-1T (liquids)
    Light source
    LED
    Temperature measurement
    5...50 °C
    Temperature sensor
    ±0,2 °C
    Temperature sensor accuracy
    ±0,2 °C
  • NAR-1T (solids)
    0 − 95 °Bx
    1,30 − 1,70 nD
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 780,00 €
    3 780,00 €
    1 * 1 ST
       3 780,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ATAG1212
    Model
    NAR-1T (solids)
    Light source
    LED
    Temperature measurement
    5...50 °C
    Temperature sensor
    ±0,2 °C
    Temperature sensor accuracy
    ±0,2 °C
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Secondary prism for NAR-1T•LO
    Secondary prism for NAR-1T•LO
    ATAGRE-1111
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Thermo display unit (DTM-1) for NAR-1T•LO, NAR-2T•LO and NAR-2T•HI
    Thermo display unit (DTM-1) for NAR-1T•LO, NAR-2T•LO and NAR-2T•HI
    ATAGRE-1165
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Lamp cable for NAR-1T•LO
    Lamp cable for NAR-1T•LO
    ATAGRE-1175
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Secondary prism for NAR-3T
    Secondary prism for NAR-3T
    ATAGRE-1204
    Log in for availability
    1 * 1 ST