Kerosene, 194…250 °C, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:64742-47-8
:
Boiling Pt:194…250 °C
Flash Pt:70 °C
Specification Test Results
- Boiling Range:194 - 250 °C
Specification Test Results
Frequently Bought Together