Order Entry
Belgium
ContactUsLinkComponent
Silver nitrate 0.1 M (0.1 N) Reag. USP volumetric solution
Silver nitrate 0.1 M (0.1 N) Reag. USP volumetric solution
Catalog # APPC186983.1211
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Silver nitrate 0.1 M (0.1 N) Reag. USP volumetric solution
Catalog # APPC186983.1211
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Presentation:
    Clear Colourless Liquid
  • Solubility:
    Miscible with water (D 20/4) 1,012
  • pH:
    4 -5
  • Titer at 20 °C:
    0,999 - 1,001
  • Traceability :
    NIST

Specification Test Results

About this item

 :
 :169.87 g/mol
EINECS:231-853-9
UN:3082
ADR:9,III
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414