Order Entry
Belgium
ContactUsLinkComponent
Ammonium iron(II) sulphate 0.1 M (0.1 N) volumetric solution
Ammonium iron(II) sulphate 0.1 M (0.1 N) volumetric solution
Catalog # APPC181369.1611
Supplier:  PanReac AppliChem
CAS Number:  
Ammonium iron(II) sulphate 0.1 M (0.1 N) volumetric solution
Catalog # APPC181369.1611
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1.000 mL
  • Seq:
    2865

Specifications

Specification Test Results

  • Presentation:
    Clear, Light green Liquid
  • Solubility:
    Miscible with water (D 20/4) 1,045
  • Titer at 20 °C:
    0,999 - 1,001
  • Traceability :
    NIST

Specification Test Results

About this item

 :
 :284.05 g/mol
UN:2796
ADR:8,II
CAS Number:10045-89-3
Formula:H₈FeN₂O₈S₂
MDL Number:MFCD00051039