Saisie de commande
Belgium
ContactUsLinkComponent
MEM non essential amino acids 100X
MEM non essential amino acids 100X
Catalog # X0557-100
Fournisseur:  Biowest
Numéro CAS:  
undefined
MEM non essential amino acids 100X
Catalog # X0557-100
Fournisseur:  Biowest
Numéro CAS:  
Spécifications
  • Cdt:
    100 ml
  • Désignation:
    MEM NEAA, 100X, without L-glutamine (liquid)
Spécifications
À propos de cet article
Concentrated solutions of amino acids, vitamins or non essential amino acids can be added to a basal medium to enrich it and the resulting medium used as a classical MEM or BME. The final concentration of the MEM or BME must be 1X.