ContactUsLinkComponent
Accessories for Memmert appliances
Accessories for Memmert appliances
Catalog # 466-5039
Supplier:  MEMMERT
Accessories for Memmert appliances
Catalog # 466-5039
Supplier:  MEMMERT
Specifications
  • Description:
    Additional blank, formatted chip card (32 kB MEMoryCard XL)
  • Pk:
    1
  • For:
    Maximum of 40 ramps, suitable for use with all Memmert units