Orders LinkContactUsLinkComponent
Multi-channel adjustable tip spacing equalizer pipettes, electronic, with bluetooth, E1-ClipTip™
Multi-channel adjustable tip spacing equalizer pipettes, electronic, with bluetooth, E1-ClipTip™
   Options durables disponibles
Fournisseur:   Thermo Fisher Scientific
Multi-channel adjustable tip spacing equalizer pipettes, electronic, with bluetooth, E1-ClipTip™
   Options durables disponibles
Certificats
Multi-channel adjustable tip spacing equalizer pipettes, electronic, with bluetooth, E1-ClipTip™
Multi-channel adjustable tip spacing equalizer pipettes, electronic, with bluetooth, E1-ClipTip™
10 Options disponibles ci-dessous
Product Details & Documents
These pipettes are ideal for quickly and efficiently performing sample transfers between virtually any tube, rack, microplate or horizontal gel box. Adjustable tip spacing allows the user to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format. Electronic tip ejection and index finger pipetting action allow the thumb to relax during pipetting.
  • ClipTip Interlocking Technology: Tip is locked and sealed in place until ejected − no dropped tips that waste valuable samples
  • Two operating options for every application: Step based Matrix or Presets with settings for most common applications
  • Five user profiles, personalised user interface and password protection offer unified operation for multiple users and environments
  • Ability to store up to 30 common protocols with a specific name and calibration in the Programs function
  • Service and calibration trackers remind users when the pipettor requires service or repair
  • Lithium-ion battery ensures long-lasting performance; unit may be recharged by a charging stand or a plug
  • Pipettes can be programmed via PC by connecting to the new My Pipette™ Creator app via Bluetooth* or USB connection
Ordering information: Supplied with universal power supply. Two-year extended warranty only available via web registration.
Caution: Please note: ClipTip™ 384 and ClipTip™ 12,5 µl tips and compatible E1-ClipTip™ models do not have interlocking technology.
* Bluetooth models are not available in all countries.
Spécifications de la famille de produits
Spécifications
  • Catalog No:
  • 613-5763
  • 613-5764
  • 613-5765
  • 613-5766
  • 613-5767
  • 613-5768
  • 613-5769
  • 613-5770
  • 613-5771
  • 613-5772
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (Certificate Registration No. EMS 623400)
  • ISO 14001 (Certificate Registration No. EMS 623400)
  • ISO 14001 (Certificate Registration No. EMS 623400)
  • ISO 14001 (Certificate Registration No. EMS 623400)
  • ISO 14001 (Certificate Registration No. EMS 623400)
  • ISO 14001 (Certificate Registration No. EMS 623400)
  • ISO 14001 (Certificate Registration No. EMS 623400)
  • ISO 14001 (Certificate Registration No. EMS 623400)
  • ISO 14001 (Certificate Registration No. EMS 623400)
  • ISO 14001 (Certificate Registration No. EMS 623400)
Spécifications
Product Family Options
6-channel
Informations sur le produit
  • Volume
    Increment
    Accuracy
    Imprecision
    Colour code
    Recommended tips
    With
    Disponibilité
    Prix
  • 15 - 1250 µl
    1 µl
    ±4,00 - ±1,44 %
    1,50 - 0,60 %
    Turquoise
    ClipTip™ 1250 µl
    9,0 - 19,8 mm adjustable tip spacing
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 1 807,00 €
    1 807,00 €
    1 * 1 ST
       1 807,00 €
    Spécifications :
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    613-5763
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No. EMS 623400)
      Low Manufacturing Impact
8-channel
Informations sur le produit
  • Volume
    Increment
    Accuracy
    Imprecision
    Colour code
    Recommended tips
    With
    Disponibilité
    Prix
  • 2 - 125 µl
    0,1 µl
    ±8,00 - ±2,00 %
    4,00 - 0,60 %
    Yellow
    ClipTip™ 200 µl
    9,0 - 14,2 mm adjustable tip spacing
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 1 960,00 €
    1 960,00 €
    1 * 1 ST
       1 960,00 €
    Spécifications :
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    613-5764
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No. EMS 623400)
      Low Manufacturing Impact
  • 10 - 300 µl
    0,1 µl
    ±5,00 - ±2,00 %
    2,00 - 0,60 %
    Orange
    ClipTip™ 300 µl
    9,0 - 14,2 mm adjustable tip spacing
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 1 960,00 €
    1 960,00 €
    1 * 1 ST
       1 960,00 €
    Spécifications :
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    613-5765
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No. EMS 623400)
      Low Manufacturing Impact
  • 15 - 1250 µl
    1 µl
    ±4,00 - ±1,44 %
    1,50 - 0,60 %
    Turquoise
    ClipTip™ 1250 µl
    9,0 - 14,2 mm adjustable tip spacing
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 1 960,00 €
    1 960,00 €
    1 * 1 ST
       1 960,00 €
    Spécifications :
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    613-5766
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No. EMS 623400)
      Low Manufacturing Impact
8-channel 384
Informations sur le produit
  • Volume
    Increment
    Accuracy
    Imprecision
    Colour code
    Recommended tips
    With
    Disponibilité
    Prix
  • 0,5 - 12,5 µl
    0,01 µl
    ±12,00 - ±2,50 %
    12,00 - 1,60 %
    Pink
    ClipTip™ 384 12,5 µl
    4,5 - 14,2 mm adjustable tip spacing
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 1 960,00 €
    1 960,00 €
    1 * 1 ST
       1 960,00 €
    Spécifications :
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    613-5767
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No. EMS 623400)
      Low Manufacturing Impact
  • 1 - 30 µl
    0,01 µl
    ±10,00 - ±2,00 %
    7,00 - 0,90 %
    Purple
    ClipTip™ 384 30 µl
    4,5 - 14,2 mm adjustable tip spacing
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 1 960,00 €
    1 960,00 €
    1 * 1 ST
       1 960,00 €
    Spécifications :
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    613-5768
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No. EMS 623400)
      Low Manufacturing Impact
  • 2 - 125 µl
    0,1 µl
    ±8,00 - ±2,00 %
    4,00 - 0,60 %
    Yellow
    ClipTip™ 384 125 µl
    4,5 - 14,2 mm adjustable tip spacing
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 1 960,00 €
    1 960,00 €
    1 * 1 ST
       1 960,00 €
    Spécifications :
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    613-5769
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No. EMS 623400)
      Low Manufacturing Impact
12-channel 384
Informations sur le produit
  • Volume
    Increment
    Accuracy
    Imprecision
    Colour code
    Recommended tips
    With
    Disponibilité
    Prix
  • 0,5 - 12,5 µl
    0,01 µl
    ±12,00 - ±2,50 %
    12,00 - 1,60 %
    Pink
    ClipTip™ 384 12,5 µl
    4,5 - 9,0 mm adjustable tip spacing
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 2 110,00 €
    2 110,00 €
    1 * 1 ST
       2 110,00 €
    Spécifications :
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    613-5770
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No. EMS 623400)
      Low Manufacturing Impact
  • 1 - 30 µl
    0,01 µl
    ±10,00 - ±2,00 %
    7,00 - 0,90 %
    Purple
    ClipTip™ 384 30 µl
    4,5 - 9,0 mm adjustable tip spacing
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 2 110,00 €
    2 110,00 €
    1 * 1 ST
       2 110,00 €
    Spécifications :
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    613-5771
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No. EMS 623400)
      Low Manufacturing Impact
  • 2 - 125 µl
    0,1 µl
    ±8,00 - ±2,00 %
    4,00 - 0,60 %
    Yellow
    ClipTip™ 384 125 µl
    4,5 - 9,0 mm adjustable tip spacing
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 2 110,00 €
    2 110,00 €
    1 * 1 ST
       2 110,00 €
    Spécifications :
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    613-5772
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No. EMS 623400)
      Low Manufacturing Impact
Accessories
Informations sur le produit
  • N° de catalogue
    For
    Disponibilité
    Prix
  • E1-Cliptip charging stand, for 3 pipettes, grey
    E1-Cliptip charging stand, for 3 pipettes, grey
    613-3883
    E1-Clip tip electronic single and multichannel pipette
    Connexion Pour la disponibilité
    1 * 1 ST - 618,00 €
Informations sur le produit
  • N° de catalogue
    For
    Disponibilité
    Prix
  • E1-Cliptip charging stand, for 1 pipette, grey
    E1-Cliptip charging stand, for 1 pipette, grey
    613-2962
    E1-Clip tip electronic single and multichannel pipette
    Connexion Pour la disponibilité
    1 * 1 ST - 307,00 €