Order Entry
Belgium
Orders LinkContactUsLinkComponent
Labels, pipe marking

Labels, pipe marking

Supplier:   Brady
Labels, pipe marking
Supplier:  Brady
Certificates
Labels, pipe marking
Labels, pipe marking

15 Options Available Below

Product Details & Documents

These pipe marking labels are made of high quality polyester and meet with GHS (Global Harmonised System) regulations for chemical substances and mixtures.

  • Promotes safety and simplifies maintenance
  • Arrows are pre-cut, and hence does not requires scissors additionally
  • Resistant to temperatures from –40 to +120 °C
  • Resistant to ultraviolet rays and conditions

The markers have a frame that contrasts with the color of the text to increase visibility.

Product Family Options

For gas

Product Information

  • Material
    Adhesive
    Colour
    Dimensions
    Packed
    Availability
    Price
  • BRDYN004112
    Pipe marking labels with GHS04 symbol
    Pipe marking labels with GHS04 symbol
    Polyester
    Black on yellow
    150×12 mm
    5 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,30 €
    12,30 €
    1 * 1 ST
       12,30 €

  • BRDYN004319
    Pipe marking labels with GHS04 symbol
    Pipe marking labels with GHS04 symbol
    Polyester
    Black on yellow
    150×12 mm
    5 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,30 €
    12,30 €
    1 * 1 ST
       12,30 €

  • BRDYN004483
    Pipe marking labels with GHS04 symbol
    Pipe marking labels with GHS04 symbol
    Polyester
    Black on yellow
    150×12 mm
    5 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,30 €
    12,30 €
    1 * 1 ST
       12,30 €

  • BRDYN004511
    Pipe marking labels with GHS02 and GHS04
    Pipe marking labels with GHS02 and GHS04
    Polyester
    Black on yellow
    150×12 mm
    5 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,30 €
    12,30 €
    1 * 1 ST
       12,30 €

  • BRDYN004522
    Pipe marking labels with GHS03 and GHS04 symbol
    Pipe marking labels with GHS03 and GHS04 symbol
    Polyester
    Black on yellow
    150×12 mm
    5 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,30 €
    12,30 €
    1 * 1 ST
       12,30 €

  • BRDYN002370
    Pipe marking labels
    Pipe marking labels
    Polyester
    Black on yellow
    250×26 mm
    4 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,50 €
    16,50 €
    1 * 1 ST
       16,50 €

  • BRDYN001991
    Pipe marking labels with GHS04 symbol
    Pipe marking labels with GHS04 symbol
    Polyester
    Blue/white
    250×26 mm
    4 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,50 €
    16,50 €
    1 * 1 ST
       16,50 €

For water

Product Information

  • Material
    Adhesive
    Colour
    Dimensions
    Packed
    Availability
    Price
  • BRDYN003252
    Pipe marking labels
    Pipe marking labels
    Polyester
    White/green
    150×12 mm
    5 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,60 €
    12,60 €
    1 * 1 ST
       12,60 €

  • BRDYN003298
    Pipe marking labels
    Pipe marking labels
    Polyester
    White/green
    150×12 mm
    5 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,60 €
    12,60 €
    1 * 1 ST
       12,60 €

  • BRDYN003331
    Pipe marking labels
    Pipe marking labels
    Polyester
    White/green
    150×12 mm
    5 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,60 €
    12,60 €
    1 * 1 ST
       12,60 €

  • BRDYN003392
    Pipe marking labels
    Pipe marking labels
    Polyester
    White/green
    250×26 mm
    4 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,50 €
    16,50 €
    1 * 1 ST
       16,50 €

For flammable liquids

Product Information

  • Material
    Adhesive
    Colour
    Dimensions
    Packed
    Availability
    Price
  • BRDYN004915
    Pipe marking labels with GHS02 and GHS07 symbol
    Pipe marking labels with GHS02 and GHS07 symbol
    Polyester
    Permanent acrylic
    Brown
    250×26 mm
    4 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,20 €
    16,20 €
    1 * 1 ST
       16,20 €

  • BRDYN005108
    Pipe marking labels with GHS02 symbol
    Pipe marking labels with GHS02 symbol
    Polyester
    Brown
    250×26 mm
    4 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,20 €
    16,20 €
    1 * 1 ST
       16,20 €

  • BRDYN005130
    GHS labels with symbols GHS02 and GHS07
    GHS labels with symbols GHS02 and GHS07
    Polyester
    Brown
    250×26 mm
    4 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,20 €
    16,20 €
    1 * 1 ST
       16,20 €

  • BRDYN005163
    GHS labels with symbols GHS06, GHS08 and GHS05
    GHS labels with symbols GHS06, GHS08 and GHS05
    Polyester
    Brown
    250×26 mm
    4 labels/card
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,20 €
    16,20 €
    1 * 1 ST
       16,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.