Osmolality standards
À propos de cet article
Osmolality standards provide ease of use when calibrating all types of osmometers. All Osmolality standards are manufactured using high purity raw materials. These products are prepared gravimetrically on a weight/weight basis. Both solute (salts) and solvent (water) are weighed on a balance calibrated by using OIML traceable weights.
- Ready to use and presented in convenient ampoules
- Extended shelf life and has low uncertainty of measurement
- Consistency of product-Independent, traceable
- Consist of 12 ampoules of 5 ml
The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.
Certifications: ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
13 Options disponibles ci-dessous
Product Details & Documents
Osmolality standards provide ease of use when calibrating all types of osmometers. All Osmolality standards are manufactured using high purity raw materials. These products are prepared gravimetrically on a weight/weight basis. Both solute (salts) and solvent (water) are weighed on a balance calibrated by using OIML traceable weights.
- Ready to use and presented in convenient ampoules
- Extended shelf life and has low uncertainty of measurement
- Consistency of product-Independent, traceable
- Consist of 12 ampoules of 5 ml
The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.
Certifications: ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
Documents
Product Family Options
Informations sur le produit
- OsmolalityPack typePk InfoPkAvailabilityPrice
- REGERE-OSM-50Osmolality standardOsmolality standard50 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-100Osmolality standardOsmolality standard100 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-200Osmolality standardOsmolality standard200 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-290Osmolality standardOsmolality standard290 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-300Osmolality standardOsmolality standard300 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-400Osmolality standardOsmolality standard400 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-500Osmolality standardOsmolality standard500 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-850Osmolality standardOsmolality standard850 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-900Osmolality standardOsmolality standard900 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-1000Osmolality standardOsmolality standard1000 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-1500Osmolality standardOsmolality standard1500 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-2000Osmolality standardOsmolality standard2000 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT204,00 €
- REGERE-OSM-3000Osmolality standardOsmolality standard3000 mOsm/kg H₂OAmpoule12×5 ml1 KITConnexion Pour la disponibilité1 * 1 KIT377,00 €