Saisie de commande
Belgium
ContactUsLinkComponent
HPLC performance kit
HPLC performance kit
Catalog # REST26394
Supplier:  Restek
undefined
HPLC performance kit
Catalog # REST26394
Supplier:  Restek
Specifications
  • Désignation:
    Performance kit for HPLC
  • Cdt:
    1
Specifications
About this item
This HPLC PEEK performance kit is a useful complement for the HPLC system.
Delivery: It includes 1 hub-cap adapter w/cap, 3 tubing clips, 1 clean cut tubing cutter, 5 PEEK finger tight fittings, 5 PEEK column plugs, 2 PEEK 90° tubing elbows, 2 PEEK 180° tubing elbows and 2 PEEK column connectors.