ContactUsLinkComponent
Beakers, quartz, tall form, with spout
Beakers, quartz, tall form, with spout
Catalog # TSLA75231
Supplier #: 75231
Beakers, quartz, tall form, with spout
Catalog # TSLA75231
Supplier #: 75231
Log in for availability
Select an Option
30 mm
Select an Option
25 ml
Select an Option
55 mm
Select an Option
02
Select an Option
1
About this item
Quartz glass, with spout.
  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature
The combination of the special thermal, electrical, chemical, mechanical and optical properties in a single material makes quartz glass indispensable for high-technology applications.
Frequently Bought Together