ContactUsLinkComponent
Shoe covers, AlphaTec® Sure Step, model 403
Shoe covers, AlphaTec® Sure Step, model 403
  ANSEWHSSS-00403-00
 :  Ansell
undefined
Shoe covers, AlphaTec® Sure Step, model 403
  ANSEWHSSS-00403-00
 :  Ansell
  • Shoe size:
    42 - 46
  • Anti-static:
    +
  • Garment application:
    General Purpose,
    Laboratory,
    Industrial
  • Colour:
    White
  • Seq:
    02
  • Model:
    403
  • Pk:
    300 PAIR
  • Fabric:
    PP film
  • Seam type:
    Stitched
  • Weight:
    65 g/m²
A disposable shoe cover that provides slip-resistance to the underside of the feet.
  • Liquid resistant material
  • Low linting
  • Anti-static
  • Elasticated opening
Weight: 65 g/m²