Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cleaning Agents
Detergents and Surface Cleansers
Cleaning solution for dissolved oxygen sensors
Cleaning solution for dissolved oxygen sensors
662-6242
:
WTW
Cleaning solution for dissolved oxygen sensors
662-6242
:
WTW
Description:
Cleaning solution for galvanic dissolved oxygen sensors StirrOx® G, CellOx® 325, DurOx® 325, TA 197 Oxi, ConOx and MPP 350 Ox module
Packed:
Bottle
Pk:
30 mL