Order Entry
Belgium
ContactUsLinkComponent
Spore discs, steel
Spore discs, steel
Catalog # NAMSTTS-06
Supplier:  CROSSTEX
undefined
Spore discs, steel
Catalog # NAMSTTS-06
Supplier:  CROSSTEX

Specifications

  • Ø:
    6 mm
  • Width:
    29 mm
  • Description:
    Geobacillus stearothermophilus, cell line 7953
  • Length:
    75 mm
  • Pk:
    100

Specifications

About this item

Spore discs are used to monitor the plasma/vaporised hydrogen peroxide sterilisation processes. Place the spore discs either directly into the chamber or positioned in areas of a device which are the most difficult to sterilise or distributed throughout rooms and isolators.

  • An inoculated carrier, with a population level of 10⁶ spores per disc
  • 19-month shelf life when stored under labelled conditions
Certifications: Meet USP and ISO 11138-1 standards.
Packaging: Packed in easy-to-access Tyvek®/Tyvek® pouches.