ContactUsLinkComponent
FlashGel™ RNA system
FlashGel™ RNA system
Supplier:  LONZA
Certificates
undefined
FlashGel™ RNA system
Supplier:  LONZA
Specifications
  • Catalog No:
  • 733-1618
  • 733-1617
  • 733-1612
  • Model:
  • FlashGel™ RNA
  • FlashGel™ RNA
  • FlashGel™ RNA
  • Gel size W×L:
  • 84×70 mm
  • 84×70 mm
  • 84×70 mm
  • Separation range (bp):
  • 0,5 kb - 9 kb
  • 0,5 kb - 9 kb
  • 0,5 kb - 9 kb
  • Storage (cassettes):
  • Room temperature for 3 months from date of manufacture
  • Room temperature for 3 months from date of manufacture
  • Room temperature for 3 months from date of manufacture
  • Well volume:
  • 2 - 5 µl
  • 2 - 5 µl
  • 2 - 5 µl
  • Cassette size W×D×H:
  • 107×115×17 mm
  • 107×115×17 mm
  • 107×115×17 mm
  • Dock size W×D×H:
  • 120×134×54 mm
  • 120×134×54 mm
  • 120×134×54 mm