Kerosene, EMPLURA®
Catalog # 1.09774.4000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:4 L
- Seq:5162
Specifications
About this item
:
Boiling Pt:180…188 °C
Flash Pt:54 °C
Storage Temperature:Ambient
EINECS:265-150-3
UN:1268
ADR:3,
:GHS02
:GHS08
CAS Number:64742-48-9