Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Urease

Urease
Supplier:  Merck
Certificates

2 Options Available Below

Product Details & Documents

Urease is used for biochemical research and analysis.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1.08489.0005
  • 1.08489.0025
  • Enzyme name:
  • Urease
  • Urease
  • Form:
  • Light yellow lyophilized powder
  • Light yellow lyophilized powder
  • Enzyme activity:
  • ≥3.0 u/mg
  • ≥3.0 u/mg
  • pH:
  • 6 - 7
  • 6 - 7

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 239,00 €
    239,00 €
    1 * 5 g
       239,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1.08489.0005
    Enzyme name
    Urease
    Form
    Light yellow lyophilized powder
    Enzyme activity
    ≥3.0 u/mg
    pH
    6 - 7
  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 800,00 €
    800,00 €
    1 * 25 g
       800,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1.08489.0025
    Enzyme name
    Urease
    Form
    Light yellow lyophilized powder
    Enzyme activity
    ≥3.0 u/mg
    pH
    6 - 7