Humidification chamber
Catalog # REST24282
Supplier: Restek
Specifications
- Description:Humidification chamber
- Material:Acrylic
- Environmentally Preferable:
- Pk:1
Specifications
About this item
It is important to use humidified air or nitrogen to help remove volatile organic contaminants when cleaning SilcoCan® or TO-Can® canisters.
- Chamber withstands pressure up to 90 psi
- Easy-to-open lid for filling with water
- Inlet and outlet compression fittings for easy connection to pressure lines on your cleaning system
It is important to use humidified air or nitrogen to help remove volatile organic contaminants when cleaning SilcoCan® or TO-Can® canisters.