Order Entry
Belgium
ContactUsLinkComponent
 
Aldehyde dehydrogenase (from Yeast)
  1.15640.0001
 :  Merck
 :  
Aldehyde dehydrogenase (from Yeast)
  1.15640.0001
 :  Merck
 :  

 

  • Pk:
    1 Vial
  • CAS No.:
    9028-88-0
  • Source:
    Yeast
  • Storage conditions:
    2...8 °C
  • Enzyme name:
    Aldehyde Dehydrogenase
  • pH:
    9,0

 

 

  • From Yeast
  • E.C.1.2.1.5
  • Freeze dried stabilized powder
  • Activity: 2 unit/mg solid (approx. 20 u/mg protein)
  • ADH: ≤0,001 U/mg
  • LDH: ≤0,001 U/mg

Definition: One unit causes the oxidation of one micromole of acetaldehyde to acetic acid per minute at pH 9,0 and 25 °C.