ContactUsLinkComponent
PID lamp polishing kit
PID lamp polishing kit
Catalog # REST20674
Supplier:  Restek
undefined
PID lamp polishing kit
Catalog # REST20674
Supplier:  Restek
Specifications
  • Description:
    Photoionisation lamp polishing kit, conatins iron oxide cleaning compound, swabs and instructions
  • Pk:
    1
Specifications
About this item
Delivery: This PID lamp polishing kit contains iron oxide cleaning compound, swabs, and instructions.