SOCO boot covers, MICROGARD® 2500, model 409
White, 42 - 46
Catalog # ANSEWH25B-00409-00
Supplier: Ansell
Jump to:
Product Details & Documents
These SOCO boot protectors are made from a non woven fabric with microporous PP film. The laminate provides an excellent barrier to chemical spray and infective agents. Developed specifically for police forensic Scene of Crime Officers (SOCOs).
- Elastic to top of boot
- Tie fastening
- Blue binding to seams
- Reinforced Surestep non slip soles
- Adjustable shoe tie
- Protection from biological agents, penetration of blood, body fluids and blood-borne pathogens
- Anti-static: Tested according to EN 1149-5
Size: One size (fits size 42 - 46)
Specifications
- Shoe size:42 - 46
- Colour:White
- Cat. No.:ANSEWH25B-00409-00
- Garment application:Medical
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.