Order Entry
Belgium
ContactUsLinkComponent
Last drop filters
Last drop filters
Catalog # REST27819
Supplier:  Restek
undefined
Last drop filters
Catalog # REST27819
Supplier:  Restek

Specifications

  • Ø:
    22,1 mm
  • Porosity:
    10 µm
  • Description:
    Last drop filter
  • Seq:
    02
  • Pk:
    1

Specifications

About this item

The flat filter element sits parallel to the bottom of the mobile phase reservoir, allowing the filter to draw 98% of the mobile phase without drawing air into the system. Conventional cylindrical mobile phase filters begin to draw air into the system when approximately 10% of the solvent remains in the reservoir.

  • Allows more analyses per batch of mobile phase
  • Helps reduce hazardous waste