Orders LinkContactUsLinkComponent
Micro extraction by packed sorbent (MEPS™)
Micro extraction by packed sorbent (MEPS™)
Micro extraction by packed sorbent (MEPS™)
Certificates
Micro extraction by packed sorbent (MEPS™)
Micro extraction by packed sorbent (MEPS™)
7 Options Available Below
Product Details & Documents
Micro Extraction by Packed Sorbent (MEPS™) is a new development in the field of sample preparation by SPE (solid phase extraction). The MEPS™ product consists of two parts, the MEPS™ syringe and the MEPS™ Barrel Insert and Needle assembly (BIN). The MEPS™ approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries. Because MEPS™ BINs can be used for sample volumes as small as 10 µl it makes them particularly well suited to on-line use with LC-MS analysis of volume limited samples.
  • The BIN contains the SPE phase that is built into the syringe needle
  • The BIN is used with a 100 or 250 µl gas tight MEPS™ syringe that allows fluid handling at normal SPE pressures
  • Sample preparation for complex biological samples is readily adapted to MEPS™ and reduces the volume of sample and reagents required for extraction
Base material is silica with mean particle size of 45 µm and pore size of 60 Å.
Product Family Options
MEPS™ syringes, removable needle
Product Information
  • Volume
    For
    Availability
    Price
  • 554-1454
    60 mm scale, 6,7 mm OD barrel
    60 mm scale, 6,7 mm OD barrel
    100 µl
    CTC Analytics, HTA 300APlus, Thermo Scientific & Varian 8400 systems
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 176,00 €
    176,00 €
    1 * 1 ST
       176,00 €
  • 549-0733
    54,1 mm scale, 6,5 mm OD barrel
    54,1 mm scale, 6,5 mm OD barrel
    100 µl
    Agilent 7693A systems
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 176,00 €
    176,00 €
    1 * 1 ST
       176,00 €
  • 554-1459
    60 mm scale, 6,7 mm OD barrel
    60 mm scale, 6,7 mm OD barrel
    250 µl
    HTA 300APlus, Thermo Scientific & Varian 8400
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 199,00 €
    199,00 €
    1 * 1 ST
       199,00 €
  • 554-1460
    60 mm scale, 7,8 mm OD barrel
    60 mm scale, 7,8 mm OD barrel
    250 µl
    CTC Analytics systems
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 199,00 €
    199,00 €
    1 * 1 ST
       199,00 €
  • 549-0734
    54,1 mm scale, 6,5 mm OD barrel
    54,1 mm scale, 6,5 mm OD barrel
    250 µl
    Agilent 7693A systems
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 199,00 €
    199,00 €
    1 * 1 ST
       199,00 €
XCHANGE® syringes for use with MEPS™ BINs
Product Information
  • Volume
    For
    Availability
    Price
  • SGEA2928500
    XCHANGE® syringe
    XCHANGE® syringe
    100 µl
    MEPS™ BINs
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €
  • SGEA2928600
    XCHANGE® syringe
    XCHANGE® syringe
    250 µl
    MEPS™ BINs
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €