Orders LinkContactUsLinkComponent
Installation kits for packed columns
Installation kits for packed columns
Installation Kit for 2 mm ID columns; for valve applications, Kit contains 1× stainless steel nut 3,2 mm, 1× stainless steel Valco nut, 1× Vespel/graphite ferrule 3,2 mm, 1× stainless steel Valco ferrule 3,2 mm, 1× stainless steel front ferrule 3,2 mm, 1× stainless steel back ferrule 3,2 mm
Catalog # REST21067
Supplier:   Restek
Installation kits for packed columns
Catalog # REST21067
Supplier:  Restek
Installation kits for packed columns
Installation kits for packed columns
Installation Kit for 2 mm ID columns; for valve applications, Kit contains 1× stainless steel nut 3,2 mm, 1× stainless steel Valco nut, 1× Vespel/graphite ferrule 3,2 mm, 1× stainless steel Valco ferrule 3,2 mm, 1× stainless steel front ferrule 3,2 mm, 1× stainless steel back ferrule 3,2 mm
Price
56,70 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications
  • Description:
    Installation Kit for 2 mm ID columns; for valve applications, Kit contains 1× stainless steel nut 3,2 mm, 1× stainless steel Valco nut, 1× Vespel/graphite ferrule 3,2 mm, 1× stainless steel Valco ferrule 3,2 mm, 1× stainless steel front ferrule 3,2 mm, 1× stainless steel back ferrule 3,2 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.