ContactUsLinkComponent
Heater/Sensor assembly
Heater/Sensor assembly
Catalog # REST27200
Supplier:  Restek
Heater/Sensor assembly
Catalog # REST27200
Supplier:  Restek
Specifications
  • Description:
    Heater/Sensor assembly
  • Pk:
    1
Specifications
About this item
Heater/Sensor assembly for Agilent 6850/6890/7890 GCs use with FID, TCD, NPD, and split/splitless inlets.